4-(Ethoxycarbonyl)-2-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Fluoro-4-ethoxycarbonylphenylboronic acid

BB-2889-1G
COBBBB-2889-1GEA 196 EUR
COBBBB-2889-1G
4-(Ethoxycarbonyl)-2-fluorphenylboronsäure
4-(Ethoxycarbonyl)-2-fluorphenylboronsäure
Formel: C₉H₁₀BFO₄
Molecular Weight: 211,9827 g/mol
MDL: MFCD08458482
CAS-Nummer: 874459-62-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR