5-Brom-2-ethoxy-3-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 5-Brom-2-ethoxypyridin-3-boronsäure, 5-Brom-2-ethoxy-3-pyridinylboronsäure

BB-2903-1G
COBBBB-2903-1GEA 520 EUR
COBBBB-2903-1G
5-Brom-2-ethoxy-3-pyridinboronsäure
5-Bromo-2-ethoxy-3-pyridineboronic acid
Formel: C₇H₉BBrNO₃
MDL: MFCD07363847
CAS-Nummer: 871332-98-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR