N-(3-Chlorpropyl)-4-boronobenzolsulfonamid

Lieferant: COMBI-BLOCKS

BB-2906-1G
COBBBB-2906-1GEA 0 EUR
COBBBB-2906-1G
N-(3-Chlorpropyl)-4-boronobenzolsulfonamid
N-(3-Chlorpropyl)-4-boronobenzolsulfonamid
Formel: C₉H₁₃BClNO₄S
MDL: MFCD08235065
CAS-Nummer: 874219-48-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR