5-Brom-2-ethoxy-4-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 5-Brom-2-ethoxypyridin-4-boronsäure, 5-Brom-2-ethoxy-4-pyridinylboronsäure

BB-2915-1G
COBBBB-2915-1GEA 303 EUR
COBBBB-2915-1G
5-Brom-2-ethoxy-4-pyridinboronsäure
5-Bromo-2-ethoxy-4-pyridineboronic acid
Formel: C₇H₉BBrNO₃
Molecular Weight: 245,87 g/mol
MDL: MFCD07783887
CAS-Nummer: 612845-46-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR