N-Benzyl-5-borono-2-methoxybenzolsulfonamid

Lieferant: COMBI-BLOCKS

Synonyme: 3-(N-Benzylsulphonamido)-4-methoxybenzeneboronic acid

BB-2920-1G
COBBBB-2920-1GEA 650 EUR
COBBBB-2920-1G
N-Benzyl-5-borono-2-methoxybenzolsulfonamid
N-Benzyl-5-borono-2-methoxybenzolsulfonamid
Formel: C₁₄H₁₆BNO₅S
MDL: MFCD08235069
CAS-Nummer: 874219-51-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR