4-(Dimethylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-3000-5G BB-3000-25G BB-3000-1G
COBBBB-3000-5GEA 0 EUR
COBBBB-3000-5G COBBBB-3000-25G COBBBB-3000-1G
4-(Dimethylcarbamoyl)phenylboronsäure
4-(Dimethylcarbamoyl)phenylboronsäure
Formel: C₉H₁₂BNO₃
MDL: MFCD02258943
CAS-Nummer: 405520-68-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR