4-(Piperidin-1-carbonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-[(Piperidin-1-yl)carbonyl]benzeneboronic acid

BB-3003-1G BB-3003-5G
COBBBB-3003-1GEA 0 EUR
COBBBB-3003-1G COBBBB-3003-5G
4-(Piperidin-1-carbonyl)phenylboronsäure
4-(Piperidin-1-carbonyl)phenylboronsäure
Formel: C₁₂H₁₆BNO₃
MDL: MFCD03411951
CAS-Nummer: 389621-83-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR