4-(4-Morpholinylcarbonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-(4-Morpholinylcarbonyl)benzeneboronic acid, (4-(Morpholine-4-carbonyl)phenyl)boronic acid

BB-3004-5G BB-3004-1G
COBBBB-3004-5GEA 0 EUR
COBBBB-3004-5G COBBBB-3004-1G
4-(4-Morpholinylcarbonyl)phenylboronsäure
4-(4-Morpholinylcarbonyl)phenylboronsäure
Formel: C₁₁H₁₄BNO₄
MDL: MFCD03411952
CAS-Nummer: 389621-84-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR