Ethyl-3-(4-boronobenzamido)propionat
Lieferant: COMBI-BLOCKS
Synonyme:
(4-((3-Ethoxy-3-oxopropyl)carbamoyl)phenyl)boronic acid, 4-[(3-Ethoxy-3-oxopropyl)carbamoyl]benzeneboronic acid
BB-3020-5G
BB-3020-1G
COBBBB-3020-5GEA
0
EUR
COBBBB-3020-5G
COBBBB-3020-1G
Ethyl-3-(4-boronobenzamido)propionat
Ethyl-3-(4-boronobenzamido)propionat
Formel:
C₁₂H₁₆BNO₅ |
MDL:
MFCD04115673 CAS-Nummer: 850568-19-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...