4-(2-Pyridylcarbamoyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
4-(Pyridin-2-yl)aminocarbonylphenylboronic acid, (4-(Pyridin-2-ylcarbamoyl)phenyl)boronic acid, 4-(2-Pyridylcarbamoyl)benzeneboronic acid
BB-3032-5G
BB-3032-1G
COBBBB-3032-5GEA
2380
EUR
COBBBB-3032-5G
COBBBB-3032-1G
4-(2-Pyridylcarbamoyl)phenylboronsäure
4-(2-Pyridylcarbamoyl)phenylboronsäure
Formel:
C₁₂H₁₁BN₂O₃ Molecular Weight: 242.04 g/mol |
MDL:
MFCD06659876 CAS-Nummer: 850568-25-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...