4-((4,5-Dihydrothiazol-2-yl)carbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (4-((4,5-Dihydrothiazol-2-yl)carbamoyl)phenyl)boronic acid

BB-3044-1G BB-3044-5G
COBBBB-3044-1GEA 0 EUR
COBBBB-3044-1G COBBBB-3044-5G
4-((4,5-Dihydrothiazol-2-yl)carbamoyl)phenylboronsäure
4-((4,5-Dihydrothiazol-2-yl)carbamoyl)phenylboronsäure
Formel: C₁₀H₁₁BN₂O₃S
MDL: MFCD06659881
CAS-Nummer: 850568-30-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR