3-(Methylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-(N-Methylaminocarbonyl)phenylboronic acid, 3-(Methylcarbamoyl)benzeneboronic acid, (3-(Methylcarbamoyl)phenyl)boronic acid

BB-3056-5G BB-3056-1G
COBBBB-3056-5GEA 0 EUR
COBBBB-3056-5G COBBBB-3056-1G
3-(Methylcarbamoyl)phenylboronsäure
3-(Methylcarbamoyl)phenylboronsäure
Formel: C₈H₁₀BNO₃
Molecular Weight: 178.98 g/mol
MDL: MFCD04038918
CAS-Nummer: 832695-88-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR