3-(Isopropylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-(N-Isopropylaminocarbonyl)benzeneboronic acid

BB-3059-1G BB-3059-5G
COBBBB-3059-1GEA 0 EUR
COBBBB-3059-1G COBBBB-3059-5G
3-(Isopropylcarbamoyl)phenylboronsäure
3-(Isopropylcarbamoyl)phenylboronsäure
Formel: C₁₀H₁₄BNO₃
MDL: MFCD04115690
CAS-Nummer: 397843-69-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR