3-(Isobutylcarbamoyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
3-(Isobutylaminocarbonyl)phenylboronic acid
BB-3061-5G
BB-3061-25G
BB-3061-1G
COBBBB-3061-5GEA
142
EUR
COBBBB-3061-5G
COBBBB-3061-25G
COBBBB-3061-1G
3-(Isobutylcarbamoyl)phenylboronsäure
3-(Isobutylcarbamoyl)phenylboronsäure
Formel:
C₁₁H₁₆BNO₃ |
MDL:
MFCD04115692 CAS-Nummer: 723282-09-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...