2-Benzyloxy-6-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-(Benzyloxy)-6-fluorophenyl)boronic acid

BB-3229-1G BB-3229-5G
COBBBB-3229-1GEA 303 EUR
COBBBB-3229-1G COBBBB-3229-5G
2-Benzyloxy-6-fluorphenylboronsäure
2-Benzyloxy-6-fluorphenylboronsäure
Formel: C₁₃H₁₂BFO₃
Molecular Weight: 246.0420 g/mol
MDL: MFCD11617256
CAS-Nummer: 1217500-53-2

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR