4-Acetyl-3-nitrophenylboronsäure

Lieferant: COMBI-BLOCKS

BB-3265-1G BB-3265-250M
COBBBB-3265-1GEA 0 EUR
COBBBB-3265-1G COBBBB-3265-250M
4-Acetyl-3-nitrophenylboronsäure
4-Acetyl-3-nitrophenylboronsäure
Formel: C₈H₈BNO₅
MDL: MFCD16618928
CAS-Nummer: 1256345-63-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR