4-(Cyclohexylcarbamoyl)-3-fluorphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(4-(Cyclohexylcarbamoyl)-3-fluorophenyl)boronic acid
BB-3307-5G
BB-3307-1G
COBBBB-3307-5GEA
1130
EUR
COBBBB-3307-5G
COBBBB-3307-1G
4-(Cyclohexylcarbamoyl)-3-fluorphenylboronsäure
4-(Cyclohexylcarbamoyl)-3-fluorphenylboronsäure
Formel:
C₁₃H₁₇BFNO₃ Molecular Weight: 265.09 g/mol |
MDL:
MFCD08436036 CAS-Nummer: 874289-11-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...