4-Carbamoyl-2-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-3318-5G BB-3318-1G
COBBBB-3318-5GEA 0 EUR
COBBBB-3318-5G COBBBB-3318-1G
4-Carbamoyl-2-fluorphenylboronsäure
4-Carbamoyl-2-fluorphenylboronsäure
Formel: C₇H₇BFNO₃
MDL: MFCD09800867
CAS-Nummer: 874289-22-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR