2-Fluor-5-(isopropylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Fluoro-5-(isopropylcarbamoyl)benzeneboronic acid, (2-Fluoro-5-(isopropylcarbamoyl)phenyl)boronic acid

BB-3344-5G BB-3344-1G
COBBBB-3344-5GEA 0 EUR
COBBBB-3344-5G COBBBB-3344-1G
2-Fluor-5-(isopropylcarbamoyl)phenylboronsäure
2-Fluor-5-(isopropylcarbamoyl)phenylboronsäure
Formel: C₁₀H₁₃BFNO₃
Molecular Weight: 225.03 g/mol
MDL: MFCD08235103
CAS-Nummer: 874289-49-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR