Benzo[b]furan-5-boronsäure

Lieferant: COMBI-BLOCKS

BB-3395-1G
COBBBB-3395-1GEA 0 EUR
COBBBB-3395-1G
Benzo[b]furan-5-boronsäure
Benzo[b]furan-5-boronsäure
Formel: C₈H₇BO₃
MDL: MFCD04038999
CAS-Nummer: 331834-13-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR