4-(2-Nitroethyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-3432-1G
COBBBB-3432-1GEA 983 EUR
COBBBB-3432-1G
4-(2-Nitroethyl)phenylboronsäure
4-(2-Nitroethyl)phenylboronsäure
Formel: C₈H₁₀BNO₄
MDL: MFCD09258738
CAS-Nummer: 957034-36-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR