N-(2-Hydroxyethyl)-3-boronobenzolsulfonamid

Lieferant: COMBI-BLOCKS

Synonyme: 3-(2-Hydroxyethylsulphamoyl)benzeneboronic acid

BB-3436-1G
COBBBB-3436-1GEA 876 EUR
COBBBB-3436-1G
N-(2-Hydroxyethyl)-3-boronobenzolsulfonamid
N-(2-Hydroxyethyl)-3-boronobenzolsulfonamid
Formel: C₈H₁₂BNO₅S
Molecular Weight: 245,06 g/mol
MDL: MFCD08436067
CAS-Nummer: 913835-57-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR