3-Fluor-4-(methoxycarbamoyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(3-Fluoro-4-(methoxycarbamoyl)phenyl)boronic acid, 3-Fluoro-4-(methoxycarbamoyl)benzeneboronic acid
BB-3438-5G
BB-3438-1G
COBBBB-3438-5GEA
1350
EUR
COBBBB-3438-5G
COBBBB-3438-1G
3-Fluor-4-(methoxycarbamoyl)phenylboronsäure
3-Fluor-4-(methoxycarbamoyl)phenylboronsäure
Formel:
C₈H₉BFNO₄ Molecular Weight: 212.97 g/mol |
MDL:
MFCD08436059 CAS-Nummer: 913835-58-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...