Imidazo[1,2-a]pyridin-6-boronsäure

Lieferant: COMBI-BLOCKS

BB-3457-1G
COBBBB-3457-1GEA 0 EUR
COBBBB-3457-1G
Imidazo[1,2-a]pyridin-6-boronsäure
Imidazo[1,2-a]pyridin-6-boronsäure
Formel: C₇H₇BN₂O₂
MDL: MFCD06739225
CAS-Nummer: 913835-63-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR