(5-(Methylthio)pyridin-3-ylthio)methylboronsäure

Lieferant: COMBI-BLOCKS

BB-3465-100M
COBBBB-3465-100MEA 742 EUR
COBBBB-3465-100M
(5-(Methylthio)pyridin-3-ylthio)methylboronsäure
(5-(Methylthio)pyridin-3-ylthio)methylboronsäure
Formel: C₇H₁₀BNO₂S₂
Molecular Weight: 215,09 g/mol
MDL: MFCD08436048
CAS-Nummer: 913835-66-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR