3-(Benzylamino)-5-nitrophenylboronsäure Hydrochlorid

Lieferant: COMBI-BLOCKS

BB-3492-1G
COBBBB-3492-1GEA 451 EUR
COBBBB-3492-1G
3-(Benzylamino)-5-nitrophenylboronsäure Hydrochlorid
3-(Benzylamino)-5-nitrophenylboronsäure Hydrochlorid
Formel: C₁₃H₁₄BClN₂O₄
MDL: MFCD08689484
CAS-Nummer: 913835-78-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR