2-Brom-6-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-Bromo-6-fluorophenyl)boronic acid, 2-Bromo-6-fluorobenzeneboronic acid

BB-3498-25G BB-3498-5G
COBBBB-3498-25GEA 121 EUR
COBBBB-3498-25G COBBBB-3498-5G
2-Brom-6-fluorphenylboronsäure
2-Brom-6-fluorphenylboronsäure
Formel: C₆H₅BBrFO₂
Molecular Weight: 218.82 g/mol
MDL: MFCD05664309
CAS-Nummer: 913835-80-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR