2-Brom-6-fluorphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(2-Bromo-6-fluorophenyl)boronic acid, 2-Bromo-6-fluorobenzeneboronic acid
BB-3498-25G
BB-3498-5G
COBBBB-3498-25GEA
121
EUR
COBBBB-3498-25G
COBBBB-3498-5G
2-Brom-6-fluorphenylboronsäure
2-Brom-6-fluorphenylboronsäure
Formel:
C₆H₅BBrFO₂ Molecular Weight: 218.82 g/mol |
MDL:
MFCD05664309 CAS-Nummer: 913835-80-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...