1-Boc-1H-pyrazol-5-boronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (1-(tert-Butoxycarbonyl)-1H-pyrazol-5-yl)boronic acid, 1H-Pyrazole-5-boronic acid, N1-BOC protected, 1-Boc-1H-pyrazole-5-boronic Acid

BB-3570-1G
COBBBB-3570-1GEA 198 EUR
COBBBB-3570-1G
1-Boc-1H-pyrazol-5-boronsäure
1-Boc-1H-pyrazole-5-boronic acid
Formel: C₈H₁₃BN₂O₄
Molecular Weight: 212,0108 g/mol
MDL: MFCD12025961
CAS-Nummer: 1217500-54-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR