2-Chlor-4-methyl-5-pyridinboronsäure

Lieferant: COMBI-BLOCKS

BB-3577-1G
COBBBB-3577-1GEA 0 EUR
COBBBB-3577-1G
2-Chlor-4-methyl-5-pyridinboronsäure
2-Chlor-4-methyl-5-pyridinboronsäure
Formel: C₆H₇BClNO₂
MDL: MFCD07368882
CAS-Nummer: 913836-08-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR