4-Brom-1-fluor-2-naphthalinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (4-Bromo-1-fluoronaphthalen-2-yl)boronic acid

BB-3583-5G BB-3583-1G
COBBBB-3583-5GEA 1180 EUR
COBBBB-3583-5G COBBBB-3583-1G
4-Brom-1-fluor-2-naphthalinboronsäure
4-Bromo-1-fluoro-2-naphthaleneboronic acid
Formel: C₁₀H₇BBrFO₂
Molecular Weight: 268.8748 g/mol
MDL: MFCD09027236
CAS-Nummer: 913836-09-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR