2,4-Difluor-5-nitrophenylboronsäure

Lieferant: COMBI-BLOCKS

BB-3589-5G BB-3589-1G
COBBBB-3589-5GEA 0 EUR
COBBBB-3589-5G COBBBB-3589-1G
2,4-Difluor-5-nitrophenylboronsäure
2,4-Difluor-5-nitrophenylboronsäure
Formel: C₆H₄BF₂NO₄
MDL: MFCD03095253
CAS-Nummer: 325786-11-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR