3-Chlor-2-fluor-4-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-Chloro-2-fluoropyridin-4-yl)boronic acid, 3-Chloro-2-fluoropyridine-4-boronic acid

BB-3591-250M
COBBBB-3591-250MEA 0 EUR
COBBBB-3591-250M
3-Chlor-2-fluor-4-pyridinboronsäure
3-Chloro-2-fluoro-4-pyridineboronic acid
Formel: C₅H₄BClFNO₂
Molecular Weight: 175,35 g/mol
MDL: MFCD09037477
CAS-Nummer: 1217500-55-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR