2,3-Difluor-5-nitrophenylboronsäure

Lieferant: COMBI-BLOCKS

BB-3801-1G BB-3801-5G
COBBBB-3801-1GEA 375 EUR
COBBBB-3801-1G COBBBB-3801-5G
2,3-Difluor-5-nitrophenylboronsäure
2,3-Difluor-5-nitrophenylboronsäure
Formel: C₆H₄BF₂NO₄
MDL: MFCD09027242
CAS-Nummer: 957060-82-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR