4-Borono-3-fluorthioanisol

Lieferant: COMBI-BLOCKS

Synonyme: 2-Fluoro-4-methylthiophenylboronic acid, (2-Fluoro-4-(methylthio)phenyl)boronic acid, 2-Fluoro-4-(methylsulphanyl)benzeneboronic acid

BB-3828-1G
COBBBB-3828-1GEA 692 EUR
COBBBB-3828-1G
4-Borono-3-fluorthioanisol
4-Borono-3-fluorthioanisol
Formel: C₇H₈BFO₂S
Molecular Weight: 186,0116 g/mol
MDL: MFCD09027247
CAS-Nummer: 957060-84-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR