2-Chlor-5-(trifluormethyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-Chlor-5-(trifluormethyl)phenyl)metaborsäure, Dihydroxy(2-chlor-5-(trifluormethyl)phenyl)boran, 2-Chlor-5-(trifluormethyl)benzolboronsäure

BB-3833-1G BB-3833-5G BB-3833-25G
COBBBB-3833-1GEA 25.5 EUR
COBBBB-3833-1G COBBBB-3833-5G COBBBB-3833-25G
2-Chlor-5-(trifluormethyl)phenylboronsäure
2-Chlor-5-(trifluormethyl)phenylboronsäure
Formel: C₇H₅BClF₃O₂
Molecular Weight: 224.37
MDL: MFCD00797335
CAS-Nummer: 182344-18-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR