3-Chlor-2-methoxy-4-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-Chloro-2-methoxypyridine-4-boronic acid, (3-Chloro-2-methoxypyridin-4-yl)boronic acid

BB-3843-1G
COBBBB-3843-1GEA 236 EUR
COBBBB-3843-1G
3-Chlor-2-methoxy-4-pyridinboronsäure
3-Chloro-2-methoxy-4-pyridineboronic acid
Formel: C₆H₇BClNO₃
Molecular Weight: 187,39 g/mol
MDL: MFCD06798255
CAS-Nummer: 957060-88-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR