5-Brom-2-methoxy-3-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 5-Brom-2-methoxy-3-pyridinylboronsäure, 5-Brom-2-methoxypyridin-3-boronsäure

BB-3860-1G
COBBBB-3860-1GEA 257 EUR
COBBBB-3860-1G
5-Brom-2-methoxy-3-pyridinboronsäure
5-Bromo-2-methoxy-3-pyridineboronic acid
Formel: C₆H₇BBrNO₃
Molecular Weight: 231,84 g/mol
MDL: MFCD04973352
CAS-Nummer: 850864-59-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR