2-Methyl-3-pyridinboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(2-Methylpyridin-3-yl)boronic acid
BB-3922-1G
COBBBB-3922-1GEA
385
EUR
COBBBB-3922-1G
2-Methyl-3-pyridinboronsäure
2-Methyl-3-pyridineboronic acid
Formel:
C₆H₈BNO₂ Molecular Weight: 136.9400 g/mol |
MDL:
MFCD07368825 CAS-Nummer: 899436-71-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...