3-(2-Nitroethyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
BB-3932-1G
COBBBB-3932-1GEA
983
EUR
COBBBB-3932-1G
3-(2-Nitroethyl)phenylboronsäure
3-(2-Nitroethyl)phenylboronsäure
Formel:
C₈H₁₀BNO₄ |
MDL:
MFCD09258757 CAS-Nummer: 957034-42-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...