2-Methyl-4-(trifluormethoxy)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Methyl-4-(trifluoromethoxy)benzeneboronic acid

BB-3945-1G
COBBBB-3945-1GEA 595 EUR
COBBBB-3945-1G
2-Methyl-4-(trifluormethoxy)phenylboronsäure
2-Methyl-4-(trifluormethoxy)phenylboronsäure
Formel: C₈H₈BF₃O₃
Molecular Weight: 219.950 g/mol
MDL: MFCD09475816
CAS-Nummer: 850033-39-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR