8-Fluor-2-methylchinolin-7-boronsäure

Lieferant: COMBI-BLOCKS

BB-3955-1G
COBBBB-3955-1GEA 1240 EUR
COBBBB-3955-1G
8-Fluor-2-methylchinolin-7-boronsäure
8-Fluor-2-methylchinolin-7-boronsäure
Formel: C₁₀H₉BFNO₂
MDL: MFCD09475865
CAS-Nummer: 957035-06-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR