5-Fluor-2-methoxy-3-pyridinboronsäure

Lieferant: COMBI-BLOCKS

BB-3971-1G
COBBBB-3971-1GEA 370 EUR
COBBBB-3971-1G
5-Fluor-2-methoxy-3-pyridinboronsäure
5-Fluoro-2-methoxy-3-pyridineboronic acid
Formel: C₆H₇BFNO₃
MDL: MFCD07368239
CAS-Nummer: 957120-32-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR