2-Chlor-3-methylphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-Chloro-3-methylphenyl)boronic acid

BB-3985-1G
COBBBB-3985-1GEA 310 EUR
COBBBB-3985-1G
2-Chlor-3-methylphenylboronsäure
2-Chlor-3-methylphenylboronsäure
Formel: C₇H₈BClO₂
Molecular Weight: 170,4000 g/mol
MDL: MFCD06797215
CAS-Nummer: 915070-53-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR