2-Chlor-6-fluor-3-methylphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-5023-1G
COBBBB-5023-1GEA 88.9 EUR
COBBBB-5023-1G
2-Chlor-6-fluor-3-methylphenylboronsäure
2-Chlor-6-fluor-3-methylphenylboronsäure
Formel: C₇H₇BClFO₂
Molecular Weight: 188,3900 g/mol
MDL: MFCD03701534
CAS-Nummer: 352535-85-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR