N-(1-Naphthyl)-4-boronobenzolsulfonamid
Lieferant: COMBI-BLOCKS
Synonyme:
(4-(N-(Naphthalen-1-yl)sulphamoyl)phenyl)boronic acid, 4-(Naphth-1-ylsulphamoyl)benzeneboronic acid
BB-5041-1G
COBBBB-5041-1GEA
312
EUR
COBBBB-5041-1G
N-(1-Naphthyl)-4-boronobenzolsulfonamid
N-(1-Naphthyl)-4-boronobenzolsulfonamid
Formel:
C₁₆H₁₄BNO₄S Molecular Weight: 327,1600 g/mol |
MDL:
MFCD09800885 CAS-Nummer: 957120-95-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...