7-Methylimidazo[1,2-a]pyridin-6-boronsäure

Lieferant: COMBI-BLOCKS

BB-5052-1G
COBBBB-5052-1GEA 0 EUR
COBBBB-5052-1G
7-Methylimidazo[1,2-a]pyridin-6-boronsäure
7-Methylimidazo[1,2-a]pyridin-6-boronsäure
Formel: C₈H₉BN₂O₂
MDL: MFCD09800890
CAS-Nummer: 957062-57-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR