3-Brom-2-fluor-5-methylphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-Bromo-2-fluoro-5-methylphenyl)boronic acid

BB-5070-5G BB-5070-1G
COBBBB-5070-5GEA 303 EUR
COBBBB-5070-5G COBBBB-5070-1G
3-Brom-2-fluor-5-methylphenylboronsäure
3-Brom-2-fluor-5-methylphenylboronsäure
Formel: C₇H₇BBrFO₂
Molecular Weight: 232.8400 g/mol
MDL: MFCD09878334
CAS-Nummer: 957066-00-1

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR