3-Hydroxy-5-(trifluormethyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-Hydroxy-5-(trifluoromethyl)phenyl)boronic acid

BB-5075-1G
COBBBB-5075-1GEA 451 EUR
COBBBB-5075-1G
3-Hydroxy-5-(trifluormethyl)phenylboronsäure
3-Hydroxy-5-(trifluormethyl)phenylboronsäure
Formel: C₇H₆BF₃O₃
Molecular Weight: 205,930 g/mol
MDL: MFCD09878337
CAS-Nummer: 957062-66-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR