4-Methoxy-3-nitrophenylboronsäure

Lieferant: COMBI-BLOCKS

BB-5076-5G BB-5076-1G
COBBBB-5076-5GEA 0 EUR
COBBBB-5076-5G COBBBB-5076-1G
4-Methoxy-3-nitrophenylboronsäure
4-Methoxy-3-nitrophenylboronsäure
Formel: C₇H₈BNO₅
MDL: MFCD03092922
CAS-Nummer: 827614-67-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR